The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145666.1 | NP_001139138.1 | 11 - 12 | 1978 | 72 |
NM_001145667.1 | NP_001139139.1 | 12 - 13 | 2011 | 72 | |
NM_012201.5 | NP_036333.2 | 12 - 13 | 2011 | 72 | |
NR_027264.1 | 12 - 13 | 2011 | 72 | ||
NR_027265.1 | 7 - 8 | 1008 | 72 | ||
GenBank mRNA | AK025457.1 | - | 9 - 10 | 1404 | 72 |
AK026972.1 | - | 3 - 4 | 370 | 72 | |
AK027032.1 | - | 12 - 13 | 1972 | 72 | |
AK304156.1 | - | 11 - 12 | 1948 | 72 | |
BC031836.1 | - | 7 - 8 | 962 | 72 | |
BC060822.1 | - | 12 - 13 | 1982 | 72 | |
U28811.1 | - | 12 - 13 | 1984 | 72 | |
U64791.1 | - | 12 - 13 | 1964 | 72 |