The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145666.1 | NP_001139138.1 | 6 - 7 | 1258 | 74 |
NM_001145667.1 | NP_001139139.1 | 7 - 8 | 1291 | 74 | |
NM_012201.5 | NP_036333.2 | 7 - 8 | 1291 | 74 | |
NR_027264.1 | 7 - 8 | 1291 | 74 | ||
GenBank mRNA | AK025457.1 | - | 4 - 5 | 684 | 74 |
AK027032.1 | - | 7 - 8 | 1252 | 74 | |
AK304156.1 | - | 6 - 7 | 1228 | 74 | |
BC060822.1 | - | 7 - 8 | 1262 | 74 | |
U28811.1 | - | 7 - 8 | 1264 | 74 | |
U64791.1 | - | 7 - 8 | 1244 | 74 |