The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286527.2 | NP_001273456.2 | 21 - 22 | 2236 | 110 |
NM_001291454.1 | NP_001278383.1 | 18 - 19 | 1816 | 110 | |
NM_014861.3 | NP_055676.3 | 21 - 22 | 2236 | 110 | |
XM_011523486.2 | XP_011521788.1 | 21 - 22 | 2326 | 110 | |
XM_011523487.2 | XP_011521789.1 | 11 - 12 | 1368 | 110 | |
GenBank mRNA | AB014603.1 | - | 19 - 20 | 2634 | 110 |
AK025424.1 | - | 6 - 7 | 684 | 110 | |
AK300526.1 | - | 21 - 22 | 2233 | 110 | |
AK316128.1 | - | 21 - 22 | 2221 | 110 | |
AY791884.1 | - | 21 - 22 | 2232 | 110 | |
BX648333.1 | - | 21 - 22 | 2233 | 110 | |
CR749829.1 | - | 21 - 22 | 2207 | 110 |