The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134647.1 | NP_001128119.1 | 15 - 16 | 2333 | 82 |
NM_198595.2 | NP_940997.1 | 14 - 15 | 2081 | 82 | |
XM_006713909.3 | XP_006713972.1 | 14 - 15 | 1964 | 82 | |
XM_011513544.2 | XP_011511846.1 | 15 - 16 | 2386 | 82 | |
XM_017008535.1 | XP_016864024.1 | 14 - 15 | 1864 | 82 | |
GenBank mRNA | AB209676.1 | - | 11 - 12 | 1534 | 82 |
AF188700.1 | - | 13 - 14 | 1808 | 82 | |
AK127962.1 | - | 6 - 7 | 1055 | 82 | |
AK290807.1 | - | 14 - 15 | 1908 | 82 | |
AK297631.1 | - | 15 - 16 | 2126 | 82 | |
BC032777.1 | - | 13 - 14 | 1832 | 82 |