Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D14
Entrez Gene ID
57533
Gene Name
TBC1 domain family member 14
Gene Aliases
-
Chromosome Location
Chr.4: 6909444 - 7033118 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113361.1 NP_001106832.1 12 - 13 1835 116
NM_001113363.1 NP_001106834.1 10 - 11 1101 116
NM_001286805.1 NP_001273734.1 10 - 11 970 116
NM_020773.2 NP_065824.2 12 - 13 1957 116
XM_005247986.2 XP_005248043.1 11 - 12 1876 116
XM_006713895.2 XP_006713958.1 12 - 13 2125 116
XM_011513507.2 XP_011511809.1 13 - 14 2044 116
XM_011513508.1 XP_011511810.1 13 - 14 1937 116
XM_011513513.2 XP_011511815.1 11 - 12 1139 116
XM_017008477.1 XP_016863966.1 11 - 12 1763 116
XM_017008478.1 XP_016863967.1 11 - 12 1207 116
XM_017008479.1 XP_016863968.1 9 - 10 971 116
XM_017008480.1 XP_016863969.1 13 - 14 1269 116
GenBank mRNA AB037743.1 - 11 - 12 1785 116
AB449900.1 - 11 - 12 1756 116
AK056962.1 - 6 - 7 1135 116
AK295068.1 - 8 - 9 715 116
AL833868.1 - 9 - 10 822 116
BC041167.1 - 12 - 13 1880 116

View Full Product Details