The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113361.1 | NP_001106832.1 | 8 - 9 | 1426 | 75 |
NM_001113363.1 | NP_001106834.1 | 6 - 7 | 692 | 75 | |
NM_001286805.1 | NP_001273734.1 | 6 - 7 | 561 | 75 | |
NM_020773.2 | NP_065824.2 | 8 - 9 | 1548 | 75 | |
XM_005247986.2 | XP_005248043.1 | 8 - 9 | 1539 | 75 | |
XM_006713895.2 | XP_006713958.1 | 8 - 9 | 1716 | 75 | |
XM_011513507.2 | XP_011511809.1 | 8 - 9 | 1533 | 75 | |
XM_011513508.1 | XP_011511810.1 | 8 - 9 | 1426 | 75 | |
XM_011513513.2 | XP_011511815.1 | 6 - 7 | 628 | 75 | |
XM_017008477.1 | XP_016863966.1 | 8 - 9 | 1426 | 75 | |
XM_017008478.1 | XP_016863967.1 | 7 - 8 | 798 | 75 | |
XM_017008479.1 | XP_016863968.1 | 6 - 7 | 634 | 75 | |
XM_017008480.1 | XP_016863969.1 | 8 - 9 | 758 | 75 | |
GenBank mRNA | AB037743.1 | - | 7 - 8 | 1376 | 75 |
AB449900.1 | - | 7 - 8 | 1347 | 75 | |
AK056962.1 | - | 3 - 4 | 798 | 75 | |
AK295068.1 | - | 3 - 4 | 204 | 75 | |
AL833868.1 | - | 5 - 6 | 413 | 75 | |
BC041167.1 | - | 8 - 9 | 1471 | 75 | |
BQ230652.1 | - | 6 - 7 | 692 | 75 |