Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIC8A
Entrez Gene ID
60626
Gene Name
RIC8 guanine nucleotide exchange factor A
Gene Aliases
RIC8
Chromosome Location
Chr.11: 207511 - 215175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286134.1 NP_001273063.1 2 - 3 1478 79
NM_021932.5 NP_068751.4 2 - 3 1478 79
XM_005253052.3 XP_005253109.1 2 - 3 664 79
XM_017018107.1 XP_016873596.1 2 - 3 748 79
GenBank mRNA AK026897.1 - 2 - 3 1477 79
AK225420.1 - 2 - 3 167 79
AK314568.1 - 2 - 3 188 79
AL136935.1 - 2 - 3 459 79
AL390088.1 - 2 - 3 199 79
BC111499.1 - 2 - 3 295 79
BC121807.1 - 2 - 3 375 79
BC121808.1 - 2 - 3 375 79

View Full Product Details