Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIC8A
Entrez Gene ID
60626
Gene Name
RIC8 guanine nucleotide exchange factor A
Gene Aliases
RIC8
Chromosome Location
Chr.11: 207511 - 215175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286134.1 NP_001273063.1 3 - 4 2069 67
NM_021932.5 NP_068751.4 3 - 4 2069 67
XM_005253052.3 XP_005253109.1 3 - 4 1255 67
XM_017018107.1 XP_016873596.1 3 - 4 1339 67
GenBank mRNA AK022870.1 - 2 - 3 623 67
AK026897.1 - 3 - 4 2068 67
AK027461.1 - 3 - 4 757 67
AK225420.1 - 3 - 4 755 67
AK314568.1 - 3 - 4 776 67
AL136935.1 - 3 - 4 1050 67
AL390088.1 - 3 - 4 790 67
BC011821.1 - 1 - 2 533 67
BC111499.1 - 3 - 4 883 67
BC121807.1 - 3 - 4 963 67
BC121808.1 - 3 - 4 963 67
BX538115.1 - 1 - 2 931 67

View Full Product Details