The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286134.1 | NP_001273063.1 | 7 - 8 | 2551 | 73 |
NM_021932.5 | NP_068751.4 | 7 - 8 | 2569 | 73 | |
XM_005253052.3 | XP_005253109.1 | 7 - 8 | 1737 | 73 | |
XM_017018107.1 | XP_016873596.1 | 7 - 8 | 1821 | 73 | |
GenBank mRNA | AK026897.1 | - | 7 - 8 | 2550 | 73 |
AK027090.1 | - | 4 - 5 | 478 | 73 | |
AK027461.1 | - | 7 - 8 | 1239 | 73 | |
AK225420.1 | - | 7 - 8 | 1237 | 73 | |
AK314568.1 | - | 7 - 8 | 1258 | 73 | |
AL136935.1 | - | 7 - 8 | 1550 | 73 | |
AL390088.1 | - | 7 - 8 | 1272 | 73 | |
BC011821.1 | - | 5 - 6 | 1015 | 73 | |
BC111499.1 | - | 7 - 8 | 1365 | 73 | |
BC121807.1 | - | 7 - 8 | 1445 | 73 | |
BC121808.1 | - | 7 - 8 | 1445 | 73 |