The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000179.2 | NP_000170.1 | 3 - 4 | 777 | 104 |
XM_011532798.1 | XP_011531100.1 | 3 - 4 | 484 | 104 | |
XM_011532799.1 | XP_011531101.1 | 3 - 4 | 559 | 104 | |
GenBank mRNA | AK308392.1 | - | 3 - 4 | 676 | 104 |
BC004246.1 | - | 3 - 4 | 677 | 104 | |
BC104665.1 | - | 3 - 4 | 699 | 104 | |
D89646.1 | - | 3 - 4 | 708 | 104 | |
U28946.1 | - | 3 - 4 | 712 | 104 | |
U54777.2 | - | 3 - 4 | 695 | 104 |