Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPYSL5
Entrez Gene ID
56896
Gene Name
dihydropyrimidinase like 5
Gene Aliases
CRAM, CRMP-5, CRMP5, Ulip6
Chromosome Location
Chr.2: 26848101 - 26950351 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253723.1 NP_001240652.1 12 - 13 1682 62
NM_001253724.1 NP_001240653.1 12 - 13 1685 62
NM_020134.3 NP_064519.2 12 - 13 1765 62
GenBank mRNA AF157634.1 - 11 - 12 1607 62
AF264015.1 - 12 - 13 1769 62
AJ251275.1 - 11 - 12 1607 62
AK022795.1 - 2 - 3 268 62
AK127314.1 - 4 - 5 2233 62
AL713706.1 - 10 - 11 1307 62
BC002874.2 - 12 - 13 1682 62
BT006871.1 - 11 - 12 1607 62

View Full Product Details