The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253723.1 | NP_001240652.1 | 8 - 9 | 1023 | 66 |
NM_001253724.1 | NP_001240653.1 | 8 - 9 | 1026 | 66 | |
NM_020134.3 | NP_064519.2 | 8 - 9 | 1106 | 66 | |
GenBank mRNA | AF157634.1 | - | 7 - 8 | 948 | 66 |
AF264015.1 | - | 8 - 9 | 1110 | 66 | |
AJ251275.1 | - | 7 - 8 | 948 | 66 | |
AL713706.1 | - | 6 - 7 | 648 | 66 | |
BC002874.2 | - | 8 - 9 | 1023 | 66 | |
BT006871.1 | - | 7 - 8 | 948 | 66 |