The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006711967.3 | XP_006712030.1 | 4 - 5 | 838 | 86 |
XM_011532700.1 | XP_011531002.1 | 4 - 5 | 631 | 86 | |
XM_017003597.1 | XP_016859086.1 | 4 - 5 | 588 | 86 | |
GenBank mRNA | BM560331.1 | - | 4 - 5 | 624 | 86 |