The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018704.2 | NP_061174.1 | 2 - 3 | 216 | 83 |
XM_011541781.2 | XP_011540083.1 | 2 - 3 | 287 | 83 | |
XM_017001806.1 | XP_016857295.1 | 2 - 3 | 654 | 83 | |
GenBank mRNA | AK022544.1 | - | 1 - 2 | 137 | 83 |
AK023340.1 | - | - | 180 | 83 | |
AL598321.1 | - | 2 - 3 | 172 | 83 | |
BC016029.1 | - | 2 - 3 | 156 | 83 |