Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDH9
Entrez Gene ID
1007
Gene Name
cadherin 9
Gene Aliases
-
Chromosome Location
Chr.5: 26880600 - 27038582 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016279.3 NP_057363.3 4 - 5 809 121
GenBank mRNA AB035302.1 - 4 - 5 794 121
AK307779.1 - 4 - 5 810 121
AK308697.1 - 4 - 5 813 121
BC107594.1 - 4 - 5 771 121
BC113745.1 - 4 - 5 771 121

View Full Product Details