The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016279.3 | NP_057363.3 | 8 - 9 | 1550 | 70 |
GenBank mRNA | AB035302.1 | - | 8 - 9 | 1535 | 70 |
AK294183.1 | - | 1 - 2 | 397 | 70 | |
AK307779.1 | - | 8 - 9 | 1551 | 70 | |
BC107594.1 | - | 8 - 9 | 1512 | 70 | |
BC113745.1 | - | 8 - 9 | 1512 | 70 |