The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006576.3 | NP_006567.3 | 2 - 3 | 169 | 72 |
XM_017018710.1 | XP_016874199.1 | 3 - 4 | 235 | 72 | |
XM_017018711.1 | XP_016874200.1 | 3 - 4 | 2981 | 72 | |
XM_017018712.1 | XP_016874201.1 | 3 - 4 | 2924 | 72 | |
XM_017018713.1 | XP_016874202.1 | 3 - 4 | 2584 | 72 | |
GenBank mRNA | AF041449.1 | - | 2 - 3 | 169 | 72 |
AK314362.1 | - | 2 - 3 | 571 | 72 | |
BC111730.1 | - | 3 - 4 | 273 | 72 |