Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AVIL
Entrez Gene ID
10677
Gene Name
advillin
Gene Aliases
ADVIL, DOC6, p92
Chromosome Location
Chr.12: 57797372 - 57818704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006576.3 NP_006567.3 9 - 10 1124 76
XM_017018710.1 XP_016874199.1 10 - 11 1190 76
XM_017018711.1 XP_016874200.1 10 - 11 3936 76
XM_017018712.1 XP_016874201.1 10 - 11 3879 76
XM_017018713.1 XP_016874202.1 10 - 11 3539 76
XM_017018714.1 XP_016874203.1 2 - 3 339 76
XM_017018715.1 XP_016874204.1 2 - 3 213 76
GenBank mRNA AF041449.1 - 9 - 10 1124 76
AK314362.1 - 9 - 10 1526 76
BC111730.1 - 10 - 11 1228 76
BX647344.1 - 1 - 2 300 76

View Full Product Details