Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFSD5
Entrez Gene ID
84975
Gene Name
major facilitator superfamily domain containing 5
Gene Aliases
hsMOT2
Chromosome Location
Chr.12: 53251251 - 53254406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170790.1 NP_001164261.1 2 - 2 1909 71
NM_032889.4 NP_116278.3 2 - 2 1636 71
XM_005269197.1 XP_005269254.1 3 - 3 2164 71
XM_005269198.4 XP_005269255.1 2 - 2 1586 71
GenBank mRNA AK097576.1 - 3 - 3 1565 71
AK225998.1 - 2 - 2 1521 71
AY358604.1 - 2 - 2 1565 71
BC007703.2 - 1 - 1 1452 71
BC067795.1 - 2 - 2 1564 71
BX640649.1 - 2 - 2 1530 71

View Full Product Details