The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000046.3 | NP_000037.2 | 3 - 4 | 1972 | 125 |
NM_198709.2 | NP_942002.1 | 4 - 5 | 1041 | 125 | |
XM_011543390.1 | XP_011541692.1 | 4 - 5 | 1185 | 125 | |
XM_011543391.2 | XP_011541693.1 | 3 - 4 | 748 | 125 | |
XM_011543392.2 | XP_011541694.1 | 3 - 4 | 748 | 125 | |
XM_011543393.1 | XP_011541695.1 | 3 - 4 | 748 | 125 | |
XM_017009471.1 | XP_016864960.1 | 3 - 4 | 748 | 125 | |
GenBank mRNA | AK290865.1 | - | 3 - 4 | 748 | 125 |
AK314903.1 | - | 3 - 4 | 757 | 125 | |
BC029051.1 | - | 4 - 5 | 1041 | 125 | |
J05225.1 | - | 3 - 4 | 692 | 125 | |
M32373.1 | - | 3 - 4 | 1245 | 125 |