Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DCTN2
Entrez Gene ID
10540
Gene Name
dynactin subunit 2
Gene Aliases
DCTN50, DYNAMITIN, HEL-S-77, RBP50
Chromosome Location
Chr.12: 57530050 - 57547331 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001261412.1 NP_001248341.1 12 - 13 1200 80
NM_001261413.1 NP_001248342.1 11 - 12 1194 80
NM_006400.4 NP_006391.1 11 - 12 1209 80
XM_011537761.2 XP_011536063.1 12 - 13 1161 80
GenBank mRNA AK055682.1 - 11 - 12 1072 80
AK096242.1 - 13 - 14 1068 80
AK098794.1 - 12 - 13 1054 80
AK222693.1 - 11 - 12 1072 80
AK292364.1 - 13 - 14 1087 80
AK314705.1 - 11 - 12 1051 80
AY189155.1 - 12 - 13 932 80
BC000718.2 - 11 - 12 1058 80
BC009468.1 - 11 - 12 984 80
BC014083.2 - 11 - 12 1032 80
FJ224315.1 - 11 - 12 1019 80
U50733.1 - 11 - 12 1019 80

View Full Product Details