The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261413.1 | NP_001248342.1 | 2 - 3 | 374 | 66 |
GenBank mRNA | AK055682.1 | - | 2 - 3 | 252 | 66 |
BC000718.2 | - | 2 - 3 | 238 | 66 | |
BC009468.1 | - | 2 - 3 | 164 | 66 | |
BC014083.2 | - | 2 - 3 | 212 | 66 | |
DB027733.1 | - | 2 - 3 | 374 | 66 |