The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002827.3 | NP_002818.1 | 2 - 3 | 361 | 140 |
GenBank mRNA | AK290638.1 | - | 2 - 3 | 328 | 140 |
AK316563.1 | - | 2 - 3 | 303 | 140 | |
AU117677.1 | - | 2 - 3 | 328 | 140 | |
BC015660.2 | - | 2 - 3 | 290 | 140 | |
BC018164.1 | - | 2 - 3 | 271 | 140 | |
BT006752.1 | - | 2 - 3 | 154 | 140 | |
DC300354.1 | - | 2 - 3 | 361 | 140 | |
M31724.1 | - | 2 - 3 | 244 | 140 | |
M33689.1 | - | 2 - 3 | 226 | 140 |