The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032849.3 | NP_116238.2 | 3 - 4 | 842 | 171 |
XM_017020801.1 | XP_016876290.1 | 2 - 3 | 1222 | 171 | |
GenBank mRNA | AB055407.1 | - | 2 - 3 | 228 | 171 |
AK027740.1 | - | - | 441 | 171 | |
BC040165.1 | - | 3 - 4 | 726 | 171 | |
BC101624.1 | - | 3 - 4 | 580 | 171 | |
BC101626.1 | - | 3 - 4 | 580 | 171 |