Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABRA2
Entrez Gene ID
2555
Gene Name
gamma-aminobutyric acid type A receptor alpha2 subunit
Gene Aliases
-
Chromosome Location
Chr.4: 46243548 - 46390039 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC022488.1 - 1 - 2 145 103

View Full Product Details