The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005023.3 | NP_005014.2 | 7 - 8 | 858 | 152 |
XM_005272020.3 | XP_005272077.2 | 7 - 8 | 873 | 152 | |
XM_011543490.2 | XP_011541792.1 | 7 - 8 | 873 | 152 | |
XM_011543492.2 | XP_011541794.1 | 7 - 8 | 870 | 152 | |
GenBank mRNA | AL520655.1 | - | 2 - 3 | 152 |