Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP57L1
Entrez Gene ID
285753
Gene Name
centrosomal protein 57 like 1
Gene Aliases
C6orf182, bA487F23.2, cep57R
Chromosome Location
Chr.6: 109095130 - 109167696 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083535.1 NP_001077004.1 11 - 12 1594 89
NM_001271852.1 NP_001258781.1 9 - 10 1443 89
NM_001271853.1 NP_001258782.1 9 - 10 1443 89
NM_173830.4 NP_776191.1 9 - 10 1209 89
XM_005266935.4 XP_005266992.3 9 - 10 1193 89
XM_005266936.4 XP_005266993.3 9 - 10 1193 89
XM_005266937.4 XP_005266994.3 9 - 10 1145 89
XM_005266938.4 XP_005266995.3 9 - 10 1142 89
XM_006715462.3 XP_006715525.1 10 - 11 1557 89
XM_011535759.2 XP_011534061.1 9 - 10 1507 89
XM_011535762.2 XP_011534064.1 7 - 8 781 89
XM_017010764.1 XP_016866253.1 10 - 11 1696 89
XM_017010765.1 XP_016866254.1 11 - 12 1951 89
XM_017010767.1 XP_016866256.1 9 - 10 1507 89
XM_017010768.1 XP_016866257.1 9 - 10 1321 89
XM_017010769.1 XP_016866258.1 9 - 10 1209 89
XM_017010770.1 XP_016866259.1 11 - 12 1900 89
XM_017010771.1 XP_016866260.1 7 - 8 786 89
XM_017010772.1 XP_016866261.1 8 - 9 1245 89
GenBank mRNA AK092723.1 - 9 - 10 1076 89
AK289910.1 - 9 - 10 1075 89
BC033448.2 - 9 - 10 1209 89
BX093163.1 - 1 - 2 134 89
DB522144.1 - 3 - 4 293 89

View Full Product Details