Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPP2R5C
Entrez Gene ID
5527
Gene Name
protein phosphatase 2 regulatory subunit B'gamma
Gene Aliases
B56G, PR61G
Chromosome Location
Chr.14: 101760573 - 101927992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161725.1 NP_001155197.1 9 - 10 989 138
NM_001161726.1 NP_001155198.1 9 - 10 1061 138
NM_002719.3 NP_002710.2 7 - 8 940 138
NM_178586.2 NP_848701.1 7 - 8 940 138
NM_178587.2 NP_848702.1 7 - 8 940 138
XM_005267819.1 XP_005267876.1 9 - 10 1050 138
XM_005267820.1 XP_005267877.1 9 - 10 990 138
XM_005267822.3 XP_005267879.1 9 - 10 1050 138
XM_005267823.1 XP_005267880.1 9 - 10 1050 138
XM_005267824.1 XP_005267881.1 9 - 10 1050 138
XM_005267826.1 XP_005267883.1 7 - 8 763 138
XM_005267827.1 XP_005267884.1 9 - 10 985 138
XM_011536914.2 XP_011535216.1 8 - 9 854 138
XM_011536915.2 XP_011535217.1 8 - 9 937 138
XM_011536916.2 XP_011535218.1 8 - 9 1936 138
XM_011536918.1 XP_011535220.1 8 - 9 1091 138
XM_011536919.2 XP_011535221.1 8 - 9 1073 138
XM_011536920.1 XP_011535222.1 8 - 9 967 138
XM_011536921.2 XP_011535223.1 7 - 8 856 138
XM_017021422.1 XP_016876911.1 8 - 9 1006 138
XM_017021423.1 XP_016876912.1 7 - 8 940 138
XM_017021424.1 XP_016876913.1 8 - 9 967 138
GenBank mRNA AB451341.1 - 7 - 8 800 138
AK131391.1 - 9 - 10 974 138
AK302470.1 - 9 - 10 889 138
AK315072.1 - 7 - 8 876 138
BC016183.1 - 7 - 8 838 138
BX248254.1 - 5 - 6 435 138
D26445.1 - 7 - 8 797 138
L42375.1 - 7 - 8 770 138
U37352.1 - 7 - 8 858 138
Z69030.1 - 7 - 8 878 138

View Full Product Details