The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080463.1 | NP_001073932.1 | 10 - 11 | 1625 | 93 |
NM_001377.2 | NP_001368.2 | 10 - 11 | 1625 | 93 | |
XM_006718903.2 | XP_006718966.1 | 10 - 11 | 1625 | 93 | |
XM_017018291.1 | XP_016873780.1 | 10 - 11 | 1625 | 93 | |
XM_017018292.1 | XP_016873781.1 | 7 - 8 | 879 | 93 | |
XM_017018293.1 | XP_016873782.1 | 10 - 11 | 1625 | 93 | |
GenBank mRNA | AB231766.1 | - | 10 - 11 | 1534 | 93 |
AB290167.1 | - | 10 - 11 | 1679 | 93 | |
AK125524.1 | - | 10 - 11 | 1625 | 93 |