The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020936.2 | NP_065987.1 | 6 - 7 | 620 | 86 |
NR_135104.1 | 6 - 7 | 620 | 86 | ||
NR_135105.1 | 6 - 7 | 620 | 86 | ||
XM_011538608.2 | XP_011536910.1 | 6 - 7 | 608 | 86 | |
XM_017019710.1 | XP_016875199.1 | 6 - 7 | 605 | 86 | |
XM_017019711.1 | XP_016875200.1 | 5 - 6 | 660 | 86 | |
XM_017019712.1 | XP_016875201.1 | 6 - 7 | 458 | 86 | |
XM_017019713.1 | XP_016875202.1 | 6 - 7 | 635 | 86 | |
XM_017019714.1 | XP_016875203.1 | 7 - 8 | 793 | 86 | |
XM_017019715.1 | XP_016875204.1 | 6 - 7 | 606 | 86 | |
XM_017019716.1 | XP_016875205.1 | 6 - 7 | 605 | 86 | |
XM_017019721.1 | XP_016875210.1 | 6 - 7 | 606 | 86 | |
GenBank mRNA | AB046815.1 | - | 1 - 2 | 2447 | 86 |
AK074219.1 | - | 6 - 7 | 572 | 86 | |
AK131442.1 | - | 3 - 4 | 859 | 86 | |
AK301043.1 | - | 6 - 7 | 583 | 86 | |
AK307384.1 | - | 6 - 7 | 596 | 86 | |
AL833756.1 | - | 4 - 5 | 1657 | 86 | |
BC030020.2 | - | 6 - 7 | 564 | 86 |