Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location
Chr.6: 152121684 - 152637399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033071.3 NP_149062.1 125 - 126 23237 96
NM_182961.3 NP_892006.3 126 - 127 23615 96
XM_006715407.1 XP_006715470.1 126 - 127 23528 96
XM_006715408.2 XP_006715471.1 126 - 127 23520 96
XM_006715409.1 XP_006715472.1 125 - 126 23507 96
XM_006715410.2 XP_006715473.1 126 - 127 23532 96
XM_006715411.1 XP_006715474.1 125 - 126 23477 96
XM_006715412.2 XP_006715475.1 126 - 127 23517 96
XM_006715413.2 XP_006715476.1 126 - 127 23532 96
XM_006715414.1 XP_006715477.1 124 - 125 23456 96
XM_006715415.2 XP_006715478.1 126 - 127 23532 96
XM_006715416.2 XP_006715479.1 126 - 127 23517 96
XM_006715417.2 XP_006715480.1 125 - 126 23391 96
XM_006715420.2 XP_006715483.1 125 - 126 23379 96
XM_006715421.2 XP_006715484.1 125 - 126 23376 96
XM_006715422.1 XP_006715485.1 125 - 126 23369 96
XM_006715423.2 XP_006715486.1 126 - 127 23532 96
XM_006715424.2 XP_006715487.1 126 - 127 23532 96
XM_006715425.2 XP_006715488.1 126 - 127 23532 96
XM_011535641.2 XP_011533943.1 126 - 127 23529 96
XM_011535642.2 XP_011533944.1 126 - 127 23517 96
XM_011535643.1 XP_011533945.1 125 - 126 23363 96
XM_011535644.1 XP_011533946.1 112 - 113 21579 96
XM_011535645.2 XP_011533947.1 108 - 109 20972 96
XM_017010608.1 XP_016866097.1 127 - 128 23647 96
XM_017010609.1 XP_016866098.1 126 - 127 23454 96
XM_017010610.1 XP_016866099.1 125 - 126 23433 96
XM_017010611.1 XP_016866100.1 125 - 126 23236 96
XM_017010612.1 XP_016866101.1 124 - 125 23160 96
XM_017010613.1 XP_016866102.1 126 - 127 23529 96
XM_017010614.1 XP_016866103.1 125 - 126 23376 96
XM_017010615.1 XP_016866104.1 125 - 126 23376 96
XM_017010616.1 XP_016866105.1 126 - 127 23532 96
XM_017010617.1 XP_016866106.1 126 - 127 23529 96
XM_017010618.1 XP_016866107.1 126 - 127 23517 96
XM_017010619.1 XP_016866108.1 112 - 113 21611 96
GenBank mRNA AB018339.2 - 14 - 15 2252 96
AF444779.1 - 1 - 2 125 96
AF495910.1 - 126 - 127 23615 96
AF535142.1 - 125 - 126 23237 96
AK122691.1 - 6 - 7 895 96
AK127076.1 - 10 - 11 1592 96
AY061755.1 - 41 - 42 6705 96
AY184206.1 - 39 - 40 6600 96
BX537837.1 - 8 - 9 1193 96
BX647837.1 - 11 - 12 1787 96

View Full Product Details