Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location
Chr.6: 152121684 - 152637399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033071.3 NP_149062.1 126 - 127 23365 62
NM_182961.3 NP_892006.3 127 - 128 23743 62
XM_006715407.1 XP_006715470.1 127 - 128 23656 62
XM_006715408.2 XP_006715471.1 127 - 128 23648 62
XM_006715409.1 XP_006715472.1 126 - 127 23635 62
XM_006715410.2 XP_006715473.1 127 - 128 23660 62
XM_006715411.1 XP_006715474.1 126 - 127 23605 62
XM_006715412.2 XP_006715475.1 127 - 128 23645 62
XM_006715413.2 XP_006715476.1 127 - 128 23660 62
XM_006715414.1 XP_006715477.1 125 - 126 23584 62
XM_006715415.2 XP_006715478.1 127 - 128 23660 62
XM_006715416.2 XP_006715479.1 127 - 128 23645 62
XM_006715417.2 XP_006715480.1 126 - 127 23519 62
XM_006715420.2 XP_006715483.1 126 - 127 23507 62
XM_006715421.2 XP_006715484.1 126 - 127 23504 62
XM_006715422.1 XP_006715485.1 126 - 127 23497 62
XM_006715423.2 XP_006715486.1 127 - 128 23660 62
XM_006715424.2 XP_006715487.1 127 - 128 23660 62
XM_006715425.2 XP_006715488.1 127 - 128 23660 62
XM_011535641.2 XP_011533943.1 127 - 128 23657 62
XM_011535642.2 XP_011533944.1 127 - 128 23645 62
XM_011535643.1 XP_011533945.1 126 - 127 23491 62
XM_011535644.1 XP_011533946.1 113 - 114 21707 62
XM_011535645.2 XP_011533947.1 109 - 110 21100 62
XM_017010608.1 XP_016866097.1 128 - 129 23775 62
XM_017010609.1 XP_016866098.1 127 - 128 23582 62
XM_017010610.1 XP_016866099.1 126 - 127 23561 62
XM_017010611.1 XP_016866100.1 126 - 127 23364 62
XM_017010612.1 XP_016866101.1 125 - 126 23288 62
XM_017010613.1 XP_016866102.1 127 - 128 23657 62
XM_017010614.1 XP_016866103.1 126 - 127 23504 62
XM_017010615.1 XP_016866104.1 126 - 127 23504 62
XM_017010616.1 XP_016866105.1 127 - 128 23660 62
XM_017010617.1 XP_016866106.1 127 - 128 23657 62
XM_017010618.1 XP_016866107.1 127 - 128 23645 62
XM_017010619.1 XP_016866108.1 113 - 114 21739 62
GenBank mRNA AB018339.2 - 15 - 16 2380 62
AF444779.1 - 2 - 3 253 62
AF495910.1 - 127 - 128 23743 62
AF535142.1 - 126 - 127 23365 62
AK122691.1 - 7 - 8 1023 62
AK127076.1 - 11 - 12 1720 62
AY061755.1 - 42 - 43 6833 62
AY184206.1 - 40 - 41 6728 62
BX537837.1 - 9 - 10 1321 62
BX647837.1 - 12 - 13 1915 62

View Full Product Details