Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location
Chr.6: 152121684 - 152637399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033071.3 NP_149062.1 142 - 143 26082 76
NM_182961.3 NP_892006.3 142 - 143 26391 76
XM_006715407.1 XP_006715470.1 144 - 145 26415 76
XM_006715408.2 XP_006715471.1 144 - 145 26407 76
XM_006715409.1 XP_006715472.1 143 - 144 26394 76
XM_006715410.2 XP_006715473.1 143 - 144 26377 76
XM_006715411.1 XP_006715474.1 143 - 144 26364 76
XM_006715412.2 XP_006715475.1 143 - 144 26362 76
XM_006715413.2 XP_006715476.1 143 - 144 26350 76
XM_006715414.1 XP_006715477.1 142 - 143 26343 76
XM_006715415.2 XP_006715478.1 142 - 143 26308 76
XM_006715416.2 XP_006715479.1 142 - 143 26293 76
XM_006715417.2 XP_006715480.1 143 - 144 26278 76
XM_006715420.2 XP_006715483.1 143 - 144 26266 76
XM_006715421.2 XP_006715484.1 143 - 144 26263 76
XM_006715422.1 XP_006715485.1 143 - 144 26256 76
XM_006715423.2 XP_006715486.1 144 - 145 26419 76
XM_006715424.2 XP_006715487.1 143 - 144 26377 76
XM_006715425.2 XP_006715488.1 142 - 143 26308 76
XM_011535641.2 XP_011533943.1 144 - 145 26416 76
XM_011535642.2 XP_011533944.1 144 - 145 26404 76
XM_011535643.1 XP_011533945.1 143 - 144 26250 76
XM_011535644.1 XP_011533946.1 130 - 131 24466 76
XM_011535645.2 XP_011533947.1 126 - 127 23859 76
XM_017010608.1 XP_016866097.1 145 - 146 26534 76
XM_017010609.1 XP_016866098.1 144 - 145 26341 76
XM_017010610.1 XP_016866099.1 143 - 144 26320 76
XM_017010611.1 XP_016866100.1 143 - 144 26123 76
XM_017010612.1 XP_016866101.1 142 - 143 26047 76
XM_017010613.1 XP_016866102.1 142 - 143 26305 76
XM_017010614.1 XP_016866103.1 143 - 144 26263 76
XM_017010615.1 XP_016866104.1 141 - 142 26152 76
XM_017010616.1 XP_016866105.1 143 - 144 26350 76
XM_017010617.1 XP_016866106.1 142 - 143 26305 76
XM_017010618.1 XP_016866107.1 142 - 143 26293 76
XM_017010619.1 XP_016866108.1 130 - 131 24498 76
GenBank mRNA AB018339.2 - 30 - 31 5028 76
AF043290.2 - 4 - 5 574 76
AF444779.1 - 17 - 18 2901 76
AF495910.1 - 142 - 143 26391 76
AF535142.1 - 142 - 143 26082 76
AK122691.1 - 22 - 23 3671 76
AK127076.1 - 27 - 28 4437 76
AK304825.1 - 16 - 17 2593 76
AK316101.1 - 15 - 16 2608 76
AL833657.1 - 7 - 8 1144 76
AY061755.1 - 57 - 58 9481 76
AY061756.1 - 15 - 16 2540 76
AY184203.1 - 14 - 15 2405 76
AY184206.1 - 55 - 56 9376 76
BC090927.1 - 15 - 16 2653 76
BX537837.1 - 24 - 25 3969 76
BX647837.1 - 28 - 29 4632 76
JQ754366.1 - 5 - 6 934 76

View Full Product Details