Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location
Chr.6: 152121684 - 152637399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033071.3 NP_149062.1 91 - 92 17767 102
NM_182961.3 NP_892006.3 92 - 93 18145 102
XM_006715407.1 XP_006715470.1 92 - 93 18043 102
XM_006715408.2 XP_006715471.1 92 - 93 18047 102
XM_006715409.1 XP_006715472.1 91 - 92 18022 102
XM_006715410.2 XP_006715473.1 92 - 93 18047 102
XM_006715411.1 XP_006715474.1 91 - 92 17992 102
XM_006715412.2 XP_006715475.1 92 - 93 18047 102
XM_006715413.2 XP_006715476.1 92 - 93 18047 102
XM_006715414.1 XP_006715477.1 90 - 91 17971 102
XM_006715415.2 XP_006715478.1 92 - 93 18047 102
XM_006715416.2 XP_006715479.1 92 - 93 18047 102
XM_006715421.2 XP_006715484.1 91 - 92 17891 102
XM_006715422.1 XP_006715485.1 91 - 92 17884 102
XM_006715423.2 XP_006715486.1 92 - 93 18047 102
XM_006715424.2 XP_006715487.1 92 - 93 18047 102
XM_006715425.2 XP_006715488.1 92 - 93 18047 102
XM_011535641.2 XP_011533943.1 92 - 93 18047 102
XM_011535642.2 XP_011533944.1 92 - 93 18047 102
XM_011535643.1 XP_011533945.1 91 - 92 17878 102
XM_011535644.1 XP_011533946.1 78 - 79 16094 102
XM_011535645.2 XP_011533947.1 74 - 75 15487 102
XM_017010608.1 XP_016866097.1 93 - 94 18162 102
XM_017010609.1 XP_016866098.1 92 - 93 17969 102
XM_017010610.1 XP_016866099.1 91 - 92 17948 102
XM_017010611.1 XP_016866100.1 91 - 92 17751 102
XM_017010612.1 XP_016866101.1 90 - 91 17675 102
XM_017010613.1 XP_016866102.1 92 - 93 18047 102
XM_017010616.1 XP_016866105.1 92 - 93 18047 102
XM_017010617.1 XP_016866106.1 92 - 93 18047 102
XM_017010618.1 XP_016866107.1 92 - 93 18047 102
XM_017010619.1 XP_016866108.1 78 - 79 16126 102
GenBank mRNA AF495910.1 - 92 - 93 18145 102
AF535142.1 - 91 - 92 17767 102
AY061755.1 - 7 - 8 1235 102
AY184206.1 - 5 - 6 1130 102

View Full Product Details