Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPN13
Entrez Gene ID
92291
Gene Name
calpain 13
Gene Aliases
-
Chromosome Location
Chr.2: 30610382 - 30809527 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144575.2 NP_653176.2 10 - 11 1267 97
XM_011533159.2 XP_011531461.1 10 - 11 1264 97
XM_011533160.2 XP_011531462.1 11 - 12 1455 97
XM_011533161.2 XP_011531463.1 10 - 11 1264 97
XM_011533163.2 XP_011531465.1 10 - 11 1264 97
XM_017005264.1 XP_016860753.1 11 - 12 1455 97
XM_017005265.1 XP_016860754.1 10 - 11 1264 97
XM_017005266.1 XP_016860755.1 10 - 11 1264 97
GenBank mRNA AK074418.1 - 11 - 12 1334 97
AK290298.1 - 10 - 11 1268 97
BC117345.1 - 10 - 11 1228 97
BC117347.1 - 10 - 11 1228 97
BX640793.1 - 10 - 11 1267 97
BX647678.1 - 3 - 4 362 97

View Full Product Details