The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278610.1 | NP_001265539.1 | 9 - 10 | 1799 | 71 |
NM_002466.3 | NP_002457.1 | 10 - 11 | 1871 | 71 | |
GenBank mRNA | AK223482.1 | - | 10 - 11 | 1606 | 71 |
AK303249.1 | - | 9 - 10 | 1704 | 71 | |
AK314791.1 | - | 10 - 11 | 1776 | 71 | |
BC007585.1 | - | 10 - 11 | 1796 | 71 | |
BC053555.1 | - | 10 - 11 | 1765 | 71 | |
X13293.1 | - | 10 - 11 | 1733 | 71 |