Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYBL2
Entrez Gene ID
4605
Gene Name
MYB proto-oncogene like 2
Gene Aliases
B-MYB, BMYB
Chromosome Location
Chr.20: 43667019 - 43716496 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278610.1 NP_001265539.1 11 - 12 2019 62
NM_002466.3 NP_002457.1 12 - 13 2091 62
GenBank mRNA AK223482.1 - 12 - 13 1826 62
AK303249.1 - 11 - 12 1924 62
AK314791.1 - 12 - 13 1996 62
BC007585.1 - 12 - 13 2016 62
BC053555.1 - 12 - 13 1985 62
BX647151.1 - 2 - 3 2476 62
X13293.1 - 12 - 13 1953 62

View Full Product Details