The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022139.3 | NP_071422.1 | 2 - 3 | 396 | 78 |
XM_005260793.2 | XP_005260850.1 | 2 - 3 | 396 | 78 | |
GenBank mRNA | AF253318.1 | - | 2 - 3 | 396 | 78 |
AJ291673.1 | - | 2 - 3 | 396 | 78 | |
AJ291675.1 | - | 2 - 3 | 396 | 78 |