Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AVIL
Entrez Gene ID
10677
Gene Name
advillin
Gene Aliases
ADVIL, DOC6, p92
Chromosome Location
Chr.12: 57797372 - 57818704 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006576.3 NP_006567.3 14 - 15 1850 78
XM_017018710.1 XP_016874199.1 15 - 16 1916 78
XM_017018711.1 XP_016874200.1 15 - 16 4662 78
XM_017018712.1 XP_016874201.1 15 - 16 4605 78
XM_017018713.1 XP_016874202.1 15 - 16 4265 78
XM_017018714.1 XP_016874203.1 7 - 8 1065 78
XM_017018715.1 XP_016874204.1 7 - 8 939 78
GenBank mRNA AF041449.1 - 14 - 15 1850 78
AK314362.1 - 14 - 15 2252 78
BC111730.1 - 15 - 16 1954 78
BX647344.1 - 5 - 6 1126 78

View Full Product Details