Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKCE
Entrez Gene ID
5581
Gene Name
protein kinase C epsilon
Gene Aliases
PKCE, nPKC-epsilon
Chromosome Location
Chr.2: 45651315 - 46187990 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005400.2 NP_005391.1 11 - 12 1791 95
XM_005264428.1 XP_005264485.1 12 - 13 1820 95
XM_006712050.3 XP_006712113.1 6 - 7 1151 95
XM_011532971.2 XP_011531273.1 13 - 14 2444 95
XM_011532975.2 XP_011531277.1 15 - 16 1932 95
XM_011532978.1 XP_011531280.1 12 - 13 1424 95
XM_011532980.2 XP_011531282.1 12 - 13 1461 95
XM_011532981.2 XP_011531283.1 10 - 11 2584 95
XM_011532982.1 XP_011531284.1 9 - 10 1596 95
XM_011532983.2 XP_011531285.1 5 - 6 2067 95
XM_017004486.1 XP_016859975.1 13 - 14 2486 95
XM_017004487.1 XP_016859976.1 13 - 14 1533 95
XM_017004488.1 XP_016859977.1 12 - 13 2268 95
XM_017004489.1 XP_016859978.1 16 - 17 1873 95
XM_017004490.1 XP_016859979.1 12 - 13 2373 95
XM_017004491.1 XP_016859980.1 11 - 12 1424 95
GenBank mRNA AK313842.1 - 11 - 12 1690 95
AL832345.1 - 11 - 12 1921 95
BC109033.2 - 11 - 12 1729 95
BC109034.2 - 11 - 12 1729 95
X65293.1 - 11 - 12 1609 95

View Full Product Details