Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKCE
Entrez Gene ID
5581
Gene Name
protein kinase C epsilon
Gene Aliases
PKCE, nPKC-epsilon
Chromosome Location
Chr.2: 45651315 - 46187990 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005400.2 NP_005391.1 4 - 5 801 66
XM_005264428.1 XP_005264485.1 5 - 6 830 66
XM_005264431.3 XP_005264488.1 4 - 5 1390 66
XM_011532971.2 XP_011531273.1 6 - 7 1454 66
XM_011532975.2 XP_011531277.1 8 - 9 942 66
XM_011532978.1 XP_011531280.1 5 - 6 434 66
XM_011532980.2 XP_011531282.1 5 - 6 471 66
XM_011532981.2 XP_011531283.1 3 - 4 1594 66
XM_011532982.1 XP_011531284.1 2 - 3 606 66
XM_017004486.1 XP_016859975.1 6 - 7 1496 66
XM_017004487.1 XP_016859976.1 6 - 7 543 66
XM_017004488.1 XP_016859977.1 5 - 6 1278 66
XM_017004489.1 XP_016859978.1 9 - 10 883 66
XM_017004490.1 XP_016859979.1 5 - 6 1383 66
XM_017004491.1 XP_016859980.1 4 - 5 434 66
XM_017004492.1 XP_016859981.1 4 - 5 1390 66
GenBank mRNA AK313842.1 - 4 - 5 700 66
AL832345.1 - 4 - 5 931 66
BC109033.2 - 4 - 5 739 66
BC109034.2 - 4 - 5 739 66
X65293.1 - 4 - 5 619 66

View Full Product Details