The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144962.2 | NP_659399.2 | 3 - 4 | 356 | 77 |
XM_011544414.2 | XP_011542716.1 | 3 - 4 | 411 | 77 | |
GenBank mRNA | AY037148.1 | - | 3 - 4 | 329 | 77 |
AY359109.1 | - | 3 - 4 | 352 | 77 | |
BC020779.1 | - | 3 - 4 | 348 | 77 | |
BF853000.1 | - | 3 - 4 | 356 | 77 | |
FJ224332.1 | - | 3 - 4 | 348 | 77 |