The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000179.2 | NP_000170.1 | 7 - 8 | 3789 | 77 |
NM_001281492.1 | NP_001268421.1 | 5 - 6 | 3399 | 77 | |
NM_001281493.1 | NP_001268422.1 | 6 - 7 | 3619 | 77 | |
NM_001281494.1 | NP_001268423.1 | 7 - 8 | 3524 | 77 | |
XM_011532798.1 | XP_011531100.1 | 7 - 8 | 3496 | 77 | |
XM_011532799.1 | XP_011531101.1 | 7 - 8 | 3571 | 77 | |
GenBank mRNA | AK130683.1 | - | 4 - 5 | 2328 | 77 |
AK293921.1 | - | 5 - 6 | 3298 | 77 | |
BC004246.1 | - | 7 - 8 | 3689 | 77 | |
BC071594.1 | - | 6 - 7 | 3622 | 77 | |
U28946.1 | - | 7 - 8 | 3724 | 77 | |
U54777.2 | - | 7 - 8 | 3707 | 77 |