Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC7
Entrez Gene ID
79741
Gene Name
coiled-coil domain containing 7
Gene Aliases
BIOT2, BioT2-A, BioT2-B, BioT2-C, C10orf68
Chromosome Location
Chr.10: 32446082 - 32882864 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321115.1 NP_001308044.1 25 - 26 3062 134
XM_005252593.2 XP_005252650.1 6 - 7 849 134
XM_006717505.1 XP_006717568.1 23 - 24 2557 134
XM_006717506.1 XP_006717569.1 22 - 23 2449 134
XM_011519663.1 XP_011517965.1 25 - 26 2770 134
XM_011519664.1 XP_011517966.1 25 - 26 2770 134
XM_011519666.1 XP_011517968.1 25 - 26 2770 134
XM_011519667.1 XP_011517969.1 25 - 26 2770 134
XM_011519668.1 XP_011517970.1 24 - 25 2680 134
XM_011519669.1 XP_011517971.1 24 - 25 2668 134
XM_011519670.1 XP_011517972.1 24 - 25 2662 134
XM_011519671.1 XP_011517973.1 25 - 26 2770 134
XM_011519672.1 XP_011517974.1 24 - 25 2659 134
XM_011519674.1 XP_011517976.1 25 - 26 2770 134
XM_011519675.1 XP_011517977.1 23 - 24 2560 134
XM_011519677.1 XP_011517979.1 24 - 25 2548 134
XM_011519679.1 XP_011517981.1 22 - 23 2239 134
XM_011519680.2 XP_011517982.1 20 - 21 1905 134
XM_011519683.2 XP_011517985.1 14 - 15 1579 134
XM_011519684.2 XP_011517986.1 25 - 26 2770 134
XM_011519685.1 XP_011517987.1 17 - 18 1787 134
XM_011519686.2 XP_011517988.1 25 - 26 2770 134
XM_011519687.1 XP_011517989.1 7 - 8 694 134
XM_017016638.1 XP_016872127.1 25 - 26 2770 134
XM_017016640.1 XP_016872129.1 25 - 26 2770 134
XM_017016641.1 XP_016872130.1 20 - 21 2199 134
XM_017016642.1 XP_016872131.1 20 - 21 2272 134
XM_017016643.1 XP_016872132.1 20 - 21 1889 134
XM_017016644.1 XP_016872133.1 25 - 26 2770 134
XM_017016645.1 XP_016872134.1 19 - 20 1813 134
XM_017016646.1 XP_016872135.1 16 - 17 1700 134
XM_017016647.1 XP_016872136.1 25 - 26 2770 134
XM_017016648.1 XP_016872137.1 12 - 13 1421 134
XM_017016649.1 XP_016872138.1 12 - 13 1361 134
XM_017016652.1 XP_016872141.1 9 - 10 824 134
XM_017016653.1 XP_016872142.1 7 - 8 734 134
GenBank mRNA AK023093.1 - 6 - 7 855 134
AK301824.1 - 3 - 4 290 134
BC125090.1 - 9 - 10 1024 134
DB026046.1 - 2 - 3 208 134

View Full Product Details