The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161344.1 | NP_001154816.1 | 16 - 17 | 1964 | 67 |
NM_001161345.1 | NP_001154817.1 | 16 - 17 | 1961 | 67 | |
NM_001161346.1 | NP_001154818.1 | 16 - 17 | 1928 | 67 | |
NM_001161347.1 | NP_001154819.1 | 14 - 15 | 1688 | 67 | |
NM_018223.2 | NP_060693.2 | 16 - 17 | 1841 | 67 | |
GenBank mRNA | AF170724.1 | - | 16 - 17 | 1970 | 67 |
AK027687.1 | - | 16 - 17 | 1922 | 67 | |
AK054917.1 | - | 2 - 3 | 869 | 67 | |
AK090948.1 | - | 3 - 4 | 937 | 67 | |
AK097671.1 | - | 14 - 15 | 1687 | 67 | |
AK302785.1 | - | 14 - 15 | 1673 | 67 | |
AK304333.1 | - | 16 - 17 | 1948 | 67 | |
AL137561.1 | - | - | 806 | 67 | |
BC012072.1 | - | 15 - 16 | 1852 | 67 |