Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD2
Entrez Gene ID
26287
Gene Name
ankyrin repeat domain 2
Gene Aliases
ARPP
Chromosome Location
Chr.10: 97572441 - 97583884 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129981.2 NP_001123453.1 7 - 8 1100 104
NM_001291218.1 NP_001278147.1 8 - 9 1199 104
NM_001291219.1 NP_001278148.1 7 - 8 1100 104
NM_020349.3 NP_065082.2 8 - 9 1199 104
GenBank mRNA AB058599.1 - 8 - 9 861 104
AJ304805.1 - 8 - 9 861 104
AJ346356.1 - 2 - 3 135 104
AJ583444.1 - 8 - 9 1199 104
AK056990.1 - 8 - 9 1141 104
AK314763.1 - 8 - 9 861 104
BC020817.1 - 7 - 8 762 104

View Full Product Details