The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000090.3 | NP_000081.1 | 17 - 18 | 1315 | 67 |
GenBank mRNA | BC028178.1 | - | 17 - 18 | 1313 | 67 |
S79877.1 | - | 4 - 5 | 197 | 67 | |
X14420.1 | - | 17 - 18 | 1300 | 67 | |
X15332.1 | - | 13 - 14 | 757 | 67 |