The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130033.1 | NP_001123505.1 | 3 - 4 | 685 | 74 |
NM_001130034.1 | NP_001123506.1 | 3 - 4 | 426 | 74 | |
NM_001130035.1 | NP_001123507.1 | 3 - 4 | 426 | 74 | |
NM_001130036.1 | NP_001123508.1 | 2 - 3 | 558 | 74 | |
NM_021200.2 | NP_067023.1 | 3 - 4 | 685 | 74 | |
XM_011545192.2 | XP_011543494.1 | 3 - 4 | 1259 | 74 | |
XM_011545193.2 | XP_011543495.1 | 3 - 4 | 1259 | 74 | |
GenBank mRNA | AB209716.1 | - | 3 - 4 | 338 | 74 |
AF081583.1 | - | 2 - 3 | 202 | 74 | |
AF093249.1 | - | 3 - 4 | 310 | 74 | |
AF100612.1 | - | 3 - 4 | 310 | 74 | |
AF101054.1 | - | 3 - 4 | 681 | 74 | |
AK289620.1 | - | 3 - 4 | 338 | 74 | |
AK289909.1 | - | 3 - 4 | 304 | 74 | |
AK290109.1 | - | 2 - 3 | 558 | 74 | |
AK296033.1 | - | 3 - 4 | 320 | 74 | |
AK301299.1 | - | 3 - 4 | 321 | 74 | |
AK314748.1 | - | 3 - 4 | 282 | 74 | |
BC008075.2 | - | 3 - 4 | 364 | 74 | |
DC305763.1 | - | 3 - 4 | 426 | 74 | |
U89715.1 | - | 3 - 4 | 681 | 74 |