The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130034.1 | NP_001123506.1 | 4 - 5 | 521 | 128 |
NM_021200.2 | NP_067023.1 | 4 - 5 | 780 | 128 | |
XM_011545192.2 | XP_011543494.1 | 4 - 5 | 1354 | 128 | |
GenBank mRNA | AF081583.1 | - | 3 - 4 | 297 | 128 |
AF100612.1 | - | 4 - 5 | 405 | 128 | |
AK301299.1 | - | 4 - 5 | 416 | 128 | |
AK314748.1 | - | 4 - 5 | 377 | 128 | |
U89715.1 | - | 4 - 5 | 776 | 128 |