The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318192.1 | NP_001305121.1 | 3 - 4 | 1077 | 71 |
NM_012450.3 | NP_036582.2 | 3 - 4 | 1077 | 71 | |
XM_011516024.2 | XP_011514326.1 | 3 - 4 | 857 | 71 | |
XM_017011963.1 | XP_016867452.1 | 3 - 4 | 857 | 71 | |
GenBank mRNA | AB208809.1 | - | 3 - 4 | 1076 | 71 |
AF169301.1 | - | 3 - 4 | 1047 | 71 | |
AK313072.1 | - | 3 - 4 | 670 | 71 | |
BC030689.2 | - | 3 - 4 | 1059 | 71 |